Check Order Status
Drum Faucets
Actual product may vary
Drum Faucets
For viscous liquids
Have Questions?
Catalog Number
Closure Type
FIS08-955
Manual
Catalog number
FIS08-955
Price (SGD)
13.00
EA
Add to cart
Closure Type:
Manual
Price (SGD)
13.00
EA
Add to cart
Gasketed to prevent leakage
Self-closing
Made of brass
Internal flame arrester
3/4 in. NPT bung
Specifications
Product Type
Drum Faucet
Closure Type
Manual
For Use With (Equipment)
For Viscous Liquids