Check Order Status
Dissecting Tissue Forceps
Actual product may vary
Dissecting Tissue Forceps
Fisherbrand Dissecting Dressing Forceps are treated to increase resistance to corrosion. Constructed from ASTM™ standard stainless steel.
Have Questions?
Catalog number
FIS08-953G
Price (SGD)
89.00
EA
Add to cart
Price (SGD)
89.00
EA
Add to cart
Fisherbrand™ Dissecting Dressing Forceps are designed to exceed perfromance expectations and manufactured to exact standards
Specialized 1x2 teeth jaws grasp tissues firmly
Provides user comfort and stability
Patterned handles
Surgical-grade stainless steel
Hand Polished
Electronically passivated for corrosion resistance
Can be sterilized by Eto/gamma radiation
Rockwell hardness (HRC): 40-45
Autoclavable up to 135°C
Latex-free
Specifications
Product Type
Tissue Dissecting Forcep
Autoclavable
Autoclavable
Certifications/Compliance
ASTM
Material
Stainless Steel
Latex Free
Yes
Tip Style
Straight, serrated, fine
Length (English)
8 in.
Length (Metric)
20.3 cm