Dissecting Jewelers Microforceps
Dissecting Jewelers Microforceps
Actual product may vary
Dissecting Jewelers Microforceps
Dissecting Jewelers Microforceps
Dissecting Jewelers Microforceps
Dissecting Jewelers Microforceps
For very precise work
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTip Style
FIS08-953FCurved, smooth
FIS08-953EStraight, smooth
Catalog number FIS08-953F
Price (SGD)
40.00
EA
Add to cart
Tip Style:
Curved, smooth
Price (SGD)
40.00
EA
Add to cart
For very precise work
  • Fisherbrand™ Dissecting Jewelers Microforceps are manufactured to exact standards and exceed perfromance expectations
  • Ultrathin, smooth tips for the finest precision work
  • Smooth handles for comfort and ease of use
  • Passivated for corrosion resistance
  • ASTM™ standard stainless steel construction
  • Latex-free
  • Can be sterilized by Eto/gamma radiation
  • Rockwell hardness (HRC): 40-45
  • Fully autoclavable
Specifications
Product TypeDissecting Forcep
AutoclavableAutoclavable
Certifications/ComplianceASTM, FDA
MaterialStainless Steel
Latex FreeYes
Tip StyleCurved, smooth
Length (English)4.5 in.
Length (Metric)11.4 cm
Unit SizeEA