Curved, Serrated or Smooth Tip Microforceps
Curved, Serrated or Smooth Tip Microforceps
Actual product may vary
Curved, Serrated or Smooth Tip Microforceps
Curved, Serrated or Smooth Tip Microforceps
Curved, Serrated or Smooth Tip Microforceps
Curved, Serrated or Smooth Tip Microforceps
Fisherbrand Curved, Serrated or Smooth Tip Microforceps are designed for high precision work. Made from ASTM™ standard (F899-84) stainless steel.
Have Questions?
Catalog NumberTip Style
FIS08-953DCurved, serrated
Catalog number FIS08-953D
Price (SGD)
137.00
EA
Add to cart
Tip Style:
Curved, serrated
Price (SGD)
137.00
EA
Add to cart
  • Fisherbrand™ Curved, Serrated or Smooth Tip Microforceps are designed for high precision work
  • Made from ASTM™ standard (F899-84) stainless steel.
  • Patterned non-slip grip handles
  • Available in serrated or smooth jaws
  • Multipurpose - Useful in educational, research or healthcare setting
  • Hand polished to mirror finish
  • Made from ASTM™ standard (F899-84) stainless steel.
  • Electronically passivated for corrosion resistance
  • Can be sterilized by Eto/gamma radiation
  • Rockwell hardness (HRC): 40-45
  • Fully Autoclavable
  • Latex-free
Specifications
Product TypeMicroforcep
AutoclavableAutoclavable
Certifications/ComplianceASTM, ISO 9001, cGMP
MaterialStainless Steel
Tip StyleCurved, serrated
Length (English)8 in.
Length (Metric)20.32 cm
Unit SizeEA