Blunt-End Dissection Scissors
Blunt-End Dissection Scissors
Actual product may vary
Blunt-End Dissection Scissors
Fisherbrand Blunt-End Dissection Scissors are multipurpose and are perfect for blunt dissection.
Have Questions?
Catalog NumberLength (English)
FIS08-9504.5 in.
Catalog number FIS08-950
Price (SGD)
31.00
EA
Add to cart
Length (English):
4.5 in.
Price (SGD)
31.00
EA
Add to cart
  • Fisherbrand™ Blunt-End Dissection Scissors
  • Our forceps are designed to exceed performance expectations -This is the Fisherbrand Advantage.
  • Multipurpose - Useful in educational, research or healthcare setting
  • Straight Blades; Blunt Point; 4.5 in.(11.4 cm)
  • Made from surgical grade stainless steel
  • Finely Finished; Highly Polished
  • Electronically passivated for corrosion resistance
  • Can be sterilized by Eto/Gamma radiation; Fully Autoclavable
  • Rockwell hardness (HRC): 46-50
  • Latex-free
  • Produced under ISO 9002, GMP, cGMP guidelines
Specifications
Product TypeDissecting Scissors
Blade StyleStraight
Length (English)4.5 in.
Length (Metric)11.43 cm
MaterialStainless Steel
For Use With (Application)Surgical, Precision
Tip StyleBlunt/Blunt
Unit SizeEA