Utility Scissors
Utility Scissors
Actual product may vary
Utility Scissors
Fisherbrand Utility Scissors are for general use.
Have Questions?
Catalog NumberLength (English)
FIS08-9454.5 in.
Catalog number FIS08-945
Price (SGD)
30.00
EA
Add to cart
Length (English):
4.5 in.
Price (SGD)
30.00
EA
Add to cart
Fisherbrand Utility Scissors are for general use.
Fisherbrand™ Utility Scissors are multipurpose.
  • Straight, ASTM™ F899-84 stainless-steel blades
  • Rockwell hardness: HRC 46-50
  • Passivated to avoid rust contamination
  • Fully autoclavable
  • Sterilizable by EtO and gamma radiation
Specifications
Product TypeUtility Scissors
Blade StyleStraight
GradeSurgical
Length (English)4.5 in.
Length (Metric)11.43 cm
Length (Metric) Blade11.4 cm
MaterialStainless Steel
AutoclavableAutoclavable
For Use With (Application)EMT, trauma, utility
Unit SizeEA