Sharp-Pointed Dissecting Scissors
Sharp-Pointed Dissecting Scissors
Actual product may vary
Sharp-Pointed Dissecting Scissors
Use these scissors for a variety of fine dissection. Perfect for educational, scientific or healthcare application. Fisherbrand™ Sharp-Pointed Dissecting Scissors are finely finished, and highly polished. Made from forged from surgical-grade stainless steel. Available in multiple styles.
Have Questions?
Catalog NumberLength (Metric)Blade Style
FIS08-94011.43 cmStraight
Catalog number FIS08-940
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
11.43 cm
Blade Style:
Straight
Use these scissors for a variety of fine dissection. Perfect for educational, scientific or healthcare application. Fisherbrand™ Sharp-Pointed Dissecting Scissors are finely finished, and highly polished. Made from forged from surgical-grade stainless steel. Available in multiple styles.
  • Fisherbrand™ Sharp-Pointed Dissecting Scissors are quality controlled per U.S
  • Government specifications
  • Narrow blades 1 in.long (25mm) cutting-edge sharp point; Perfect for fine dissection
  • Electronically passivated
  • Autoclavable up to 135°C
  • Surgical grade tainless steel
  • Can be sterilized by Eto/Gamma radiation
  • Rockwell Hardness (HRC): 46-50
  • Inspected per U.S
  • Government specifications for surgical instruments
  • Produced under ISO 9001, cGMP Guidelines
Specifications
Product TypeDissecting Scissors
Blade StyleStraight
GradeSurgical
Length (English)4.5 in.
Length (Metric)11.43 cm
Length (Metric) Blade11.4 cm
MaterialStainless Steel
AutoclavableAutoclavable
For Use With (Application)Dissecting
Tip StyleSharp/Sharp
Unit SizeEA