▸
Surgical and Dissection Equipment
▸
Scalpels
▸
Single-Use Scalpels
Actual product may vary
Graham-Field
Single-Use Scalpels
Surgical quality — also ideal for student or general lab use
Have Questions?
Change view
Catalog Number
Size
FIS08-927-5A
# 10
FIS08-927-5D
# 21
2 Options
Catalog number
FIS08-927-5A
Price (SGD)
51.00
Pack
Add to cart
Size:
# 10
Price (SGD)
51.00
Pack
Add to cart
Stainless-steel blade attached to plastic handle
Sterile, individually packaged
Plastic guard protects cutting edge and prevents package puncture
Specifications
Blade Material
Stainless Steel
For Use With (Application)
Student or General Lab Use
Size
# 10
Description
Single-use Scalpel
Disposable
Single-use
Sterility
Sterile
Handle Material
Plastic