Dissection Scalpel
Dissection Scalpel
Actual product may vary
Dissection Scalpel
Dissection Scalpel
Dissection Scalpel
Fisherbrand Dissection Scalpel is made from ASTM™ standard pure nickel.
Have Questions?
Catalog NumberBlade Material
FIS08-925Nickel-plated Metal
Catalog number FIS08-925
Price (SGD)
35.00
EA
Add to cart
Blade Material:
Nickel-plated Metal
Price (SGD)
35.00
EA
Add to cart
  • Fisherbrand™ Dissection Scalpel is made from ASTM™ standard pure nickel.
  • Easy to clean and sterilization
  • Retains good cutting edge
  • Nonsterile
  • Highly polished
  • Made from ASTM™ Pure Nickel Sheet
  • Cut-Edge Length: 1.5 in.
  • Total Length: 5 in.
Specifications
Blade MaterialNickel-plated Metal
Product TypeDissecting Scalpel