Stainless-Steel Blades for Dissecting Knife Handles
Actual product may vary
Graham-Field
Stainless-Steel Blades for Dissecting Knife Handles
Exceptionally strong and uniformly sharp
Have Questions?
Catalog NumberSizeFor Use With (Equipment)Length (English)
FIS08-916-5B# 11For Handle # 3, # 5, # 71.56 in.
Catalog number FIS08-916-5B
Price (SGD)
86.00
Pack
Size:
# 11
For Use With (Equipment):
For Handle # 3, # 5, # 7
Length (English):
1.56 in.
Price (SGD)
86.00
Pack
Exceptionally strong and uniformly sharp
  • Surgical quality
  • Sterile
  • Individually foil-wrapped
Specifications
Blade MaterialStainless Steel
Product TypeBlade
Size# 11
Length (English)1.56 in.
Length (Metric)4 cm
For Use With (Equipment)For Handle # 3, # 5, # 7