Bard-Parker™ Surgical Blade Handles
Actual product may vary
Aspen Surgical
Bard-Parker™ Surgical Blade Handles
Have Questions?
Catalog NumberSize
FIS08-915-2# 3
Catalog number FIS08-915-2
Price (SGD)
95.00
Each
Size:
# 3
Price (SGD)
95.00
Each
  • Handles are designed for a contoured, firm grip and easy manipulation
  • Distal ends are tapered and beveled for blunt dissection use
Specifications
Blade MaterialCarbon Steel
Product TypeContoured Blade
Size# 3
For Use With (Equipment)# 10 to # 15 Blade for General Surgery like Tissue Separation and other Procedures that require a Sharp Surgical Blade to Puncture or Cut
DescriptionFirm Grip Blade, Tapered and Beveled Distal ends, Surgical Scalpel Handle