Dissecting Fine-Pointed Forceps
Dissecting Fine-Pointed Forceps
Actual product may vary
Dissecting Fine-Pointed Forceps
Fisherbrand Dissecting Fine-Pointed Forceps are used for fine dissection. Fine, serrated tips with guide pins.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberShape
FIS08-880Straight
FIS08-875Curved
Catalog number FIS08-880
Price (SGD)
23.00
EA
Add to cart
Shape:
Straight
Price (SGD)
23.00
EA
Add to cart
Fisherbrand Dissecting Fine-Pointed Forceps are used for fine dissection. Fine, serrated tips with guide pins.
  • Fisherbrand™ Dissecting Fine-Pointed Forceps are designed to exceed perfromance expectations and manufactured to exact standards
  • Serrated tips for grasping tissue and materials
  • Guide pins allows tips to meet precisely
  • Ribbed handles for secure grip, comfort and ease of use
  • Passivated for corrosion resistance
  • Made from ASTM™ standard stainless steel construction
  • Latex-free
  • Can be sterilized by Eto/gamma radiation
  • Rockwell hardness (HRC): 40-45
  • Fully autoclavable
Specifications
Product TypeDissecting Forcep
AutoclavableAutoclavable
Certifications/ComplianceASTM™ Standard
ColorSterling Silver
MaterialStainless Steel
FinishHigh Polish
For Use With (Application)Fine Tissue Dissection
IncludesGuide pins
Latex FreeYes
ShapeStraight
Tip StyleSerrated, fine point
Length (English)4.5 in.
Length (Metric)11.4 cm
Unit SizeEA