▸
Surgical and Dissection Equipment
▸
Dissection Kits
▸
8-Piece Dissection Kit
Actual product may vary
Fisherbrand
8-Piece Dissection Kit
Fisherbrand 8 Piece Dissection Set components are made from ASTM™ standard steel. All instruments come pre-assembled in a beautiful hard plastic case.
Have Questions?
Catalog Number
Quantity
FIS08-855
8 Pc.
Catalog number
FIS08-855
Price (SGD)
28.00
Each
Add to cart
Quantity:
8 Pc.
Price (SGD)
28.00
Each
Add to cart
Fisherbrand™ 8 Piece Dissection Set Includes:
Forceps: medium points, nickel-plated
Scissors: 1.5 in. (40mm) blades, stainless steel
Dissecting knife handle: nickel-plated
Knife blades: stainless steel, pack of 3
Dropping pipet: glass
Bent dissecting needle: stainless steel with plastic handle
Straight dissecting needle: stainless steel with plastic handle
Vinylite™ Ruler 6 in. (15cm)
Specifications
Product Type
Dissecting Set
Includes
Forcep, scissors, dissecting knife handle, knife blades, dropping pipet, bent dissecting needle, straight dissecting needle, 6 in. Vinylite™ ruler
Quantity
8 Pc.