Glass Staining Dish for 10 Slides, with Cap
Glass Staining Dish for 10 Slides, with Cap
Actual product may vary
Glass Staining Dish for 10 Slides, with Cap
Glass Staining Dish for 10 Slides, with Cap
Glass Staining Dish for 10 Slides, with Cap
Hold up to 10 standard 3 x 1 in. (75 x 25mm) slides back-to-back
Have Questions?
Catalog NumberSlide Count
FIS08-81610
Catalog number FIS08-816
Price (SGD)
42.00
EA
Add to cart
Slide Count:
10
Price (SGD)
42.00
EA
Add to cart
Hold up to 10 standard 3 x 1 in. (75 x 25mm) slides back-to-back
  • Heavy molded glass Coplin staining dish holds slides vertically
  • Rectangular body with five grooves
  • With tight-fitting plastic screw cap 
Specifications
Closure TypeScrew
MaterialGlass
TypeCoplin Staining Dish
Slide Count10
Height (Metric)11 cm
Height (English)4.31 in.
Unit SizeEA