Wheaton™ Glass Staining Dish for 5 or 10 Slides
Wheaton™ Glass Staining Dish for 5 or 10 Slides
Actual product may vary
DWK Life Sciences
Wheaton™ Glass Staining Dish for 5 or 10 Slides
For fully immersed slides
Have Questions?
Catalog NumberSlide Count
FIS08-813E5 Vertically or 10 Back to Back
Catalog number FIS08-813E
Price (SGD)
108.00
EA
Add to cart
Slide Count:
5 Vertically or 10 Back to Back
Price (SGD)
108.00
EA
Add to cart
  • Coplin staining jar holds up to five single slides or ten slides 3 × 1 in. (75 × 25mm) back-to-back vertically
  • 08-813E: Dish made from soda-lime glass
  • 02-912-618: Replacement cap: polypropylene
Specifications
Closure ColorWhite
Closure IncludedClosure Included
Closure MaterialPolypropylene
Closure TypeScrew
Bottom ShapeRectangular
Capacity (Metric)60 mL
MaterialSoda Lime Glass
TypeSlide Staining Jar
Slide Count5 Vertically or 10 Back to Back
Dimensions (L x W)1.02 x 1.02 in. (2.6 x 2.6 cm)
Length (English)1.02 in.
Length (Metric)26 mm
Width (English)1.02 in.
Width (Metric)26 mm
Depth (English) Inside3.5 in.
Depth (Metric)90 mm
Depth (Metric) Inside9 cm
Unit SizeEA