Wheaton™ Glass 20-Slide Staining Dish with Removable Rack
Wheaton™ Glass 20-Slide Staining Dish with Removable Rack
Actual product may vary
DWK Life Sciences
Wheaton™ Glass 20-Slide Staining Dish with Removable Rack
Complete with dish, cover, and glass slide rack
Have Questions?
Catalog NumberSlide Count
FIS08-81220
Catalog number FIS08-812
Price (SGD)
354.00
EA
Add to cart
Slide Count:
20
Price (SGD)
354.00
EA
Add to cart
  • With removable rack
  • Removable glass rack has a spring wire handle for manual staining of up to twenty horizontal slides
  • Bottom of rack is open for rapid immersion in and draining of stain
  • Holds slides measuring 3 × 1 in. (75 × 25mm), 3 × 1 1/2 in. (75 × 38mm), and 3 × 2 in. (75 × 51mm)
  • Made from soda-lime glass
Specifications
Bottom ShapeRectangular
MaterialSoda Lime Glass
TypeStaining Dish
IncludesRemovable Glass Slide Slotted Rack with Open Bottom
Slide Count20
Dimensions (L x W)3.7 x 2.9 in. (9.5 x 7.6 cm)
Length (English)3.7 in.
Length (Metric)95 mm
Width (English)2.9 in.
Width (Metric)76 mm
Depth (English) Inside2.5 in.
Depth (Metric)64 mm
Depth (Metric) Inside6.4 cm
Unit SizeEA