Check Order Status
Drum Lifter
Actual product may vary
Drum Lifter
Can be used with a forklift or overhead pulley
Have Questions?
Catalog Number
Material
FIS08-801
Steel
Catalog number
FIS08-801
Price (SGD)
395.00
PK
Add to cart
Material:
Steel
Price (SGD)
395.00
PK
Add to cart
Sturdy heavy-gauge steel drum lifter attaches securely to any drum rim
Holds up to 1000 lb. (454kg)
Specifications
Product Type
Tong Style Drum Lifter
For Use With (Application)
Allowing a 55 gal. Steel drum to be lifted and moved with a forklift truck
For Use With (Equipment)
Forklift or Overhead Pulley
Material
Steel