Standard Tissue Culture Dishes
Standard Tissue Culture Dishes
Actual product may vary
Standard Tissue Culture Dishes
Standard Tissue Culture Dishes
Standard Tissue Culture Dishes
Standard Tissue Culture Dishes
Constructed of flat, optically clear polystyrene surfaces for distortion-free microscopic visualization of cells. Falcon™ Standard Tissue Culture Dishes works with most attachment-dependent cell lines.
Have Questions?
Catalog NumberCulture AreaVolume (Metric) Working
FIS08-772E58.1 cm216 to 17.5 mL
Catalog number FIS08-772E
Price (SGD)
1,116.00
CS
Add to cart
Culture Area:
58.1 cm2
Volume (Metric) Working:
16 to 17.5 mL
Price (SGD)
1,116.00
CS
Add to cart
  • Flat, optically clear polystyrene surfaces for distortion-free microscopic visualization of cells
  • Uniform surface chemistry created by vacuum-gas plasma treatment promotes cell attachment
  • Improved handling of small dishes with the unique Easy-Grip design
  • Nonpyrogenic
  • Sterilized by gamma irradiation
  • Packaged in peel-open, medical-style bags
  • Crystal-grade virgin polystyrene
  • Standard Tissue Culture (TC), Corning™ PrimariaTM surface treatment and non-TC-treated polystyrene available
Designed for cell culture
  • Flat, distortion-free optics
  • Lids designed for optimal gas exchange
  • Stacking rings allow for easier stacking and handling
  • Vacuum-gas plasma treatment permanently and consistently modifies the cell growth surface
  • Standard tissue culture surface is hydrophilic and contains a variety of negatively charged functional groups that support cell attachment and spreading
  • Corning Primaria tissue culture surface additionally incorporates nitrogen-containing functional groups and has been shown to support improved attachment, spreading, and differentiation of some cell types
Specifications
Bottom ShapeFlat
Capacity (Metric)17.5 mL (Well)
ColorClear
Culture Area58.1 cm2
MaterialPolystyrene
ShapeRound
SterilitySterile
Surface TreatmentCell Culture
TypeCell Culture Petri Dish
Volume (Metric) Working16 to 17.5 mL
No. per Case200/Cs.
No. per Pack20/Pk.
Height (Metric)20 mm
Diameter (Metric)100 mm
Unit SizeCS