Polystyrene Microplates
Polystyrene Microplates
Actual product may vary
Polystyrene Microplates
Constructed of crystal grade virgin polystyrene for optimum visibility. Falcon™ Polystyrene Microplates are available with tissue culture-treated and nontreated surfaces.
Have Questions?
Catalog NumberCulture Area
FIS08-772-12 cm2
Catalog number FIS08-772-1
Price (SGD)
613.00
CS
Add to cart
Culture Area:
2 cm2
Price (SGD)
613.00
CS
Add to cart
Available with tissue culture-treated and nontreated surfaces. Made of crystal grade virgin polystyrene for optimum visibility.
  • Versatility of plate colors to suit various detection methods
  • Crystal-grade virgin polystyrene
  • Sterilized by gamma irradiation
  • Nonpyrogenic
  • Non-treated polystyrene, standard tissue culture (TC), and Corning™ PrimariaTM surface treatments available
  • Convenient, peel-open medical-style packaging for individual product presentation
  • Individual and Ready-Stack (RS) trays are PET (Code 1) and recyclable
  • Growth area and volume are nominal
Specifications
Array4 x 6
Closure ColorClear
Closure IncludedYes
Closure MaterialPolystyrene
Bottom ShapeFlat
Capacity (Metric)3.5 mL
ColorClear
Culture Area2 cm2
MaterialPolystyrene
ShapeRectangular
SterilitySterile
Surface TreatmentCell Culture
TypeCell Culture Plate
Volume (Metric) Working1.5 to 2.2 mL
Well Count24
IncludesLid
No. per Case50/Cs.
No. per Pack1/Pk.
PackagingIndividually Wrapped
Pore Size1 μm
Quantity50/Cs.
Unit SizeCS