Polystyrene Antistatic Weighing Dishes
Actual product may vary
Fisherbrand
Polystyrene Antistatic Weighing Dishes
Disposable dishes are ideal for weighing, dispensing, or storing applications
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)ColorType
FIS08-732-118270 mLNaturalPour Boat
FIS08-732-114100 mLBlueWeighing Dish
FIS08-732-1127 mLNaturalWeighing Dish
FIS08-732-11730 mLNaturalPour Boat
FIS08-732-11610 mLNaturalPour Boat
FIS08-732-115290 mLNaturalWeighing Dish
FIS08-732-113100 mLNaturalWeighing Dish
Catalog number FIS08-732-118
Price (SGD)
46.00
Case
Capacity (Metric):
270 mL
Color:
Natural
Type:
Pour Boat
Price (SGD)
46.00
Case
Fisherbrand™ Polystyrene Antistatic Weighing Dishes are contaminant-free, antistatic, and biologically inert. Disposable dishes are ideal for weighing, dispensing, or storing applications.
Ideal for weighing static-affected materials.
  • Resist dilute acids, aqueous solutions, alcohols, and bases
  • Flat bottoms resist tipping
  • Made from 12mil thick polystyrene—20% thicker than most dishes on the market today—for better strength, stability, and handling ease
  • Pour boats feature a molded spout for convenient pouring and dispensing
Specifications
Bottom ShapeFlat
Capacity (Metric)270 mL
ColorNatural
DisposableSingle-use
MaterialPolystyrene
ShapeFlat
TypePour Boat
For Use With (Application)Ideal for weighing, dispensing or storing applications
Thickness12 mil
Dimensions (L x W x H)4.75 x 7.75 x 1 in.
Length (English)4.75 in.
Length (Metric)120 mm
Width (English)7.75 in.
Width (Metric)196 mm
Height (Metric)25.4 mm
Height (English)1 in.
Diameter (Metric) Inner Base98 mm
Diameter (Metric)98 mm (Inner Base)
Unit SizeCase