Universal Dialysis Tubing Closures (for all Membrane Types)
Actual product may vary
Spectrum
Universal Dialysis Tubing Closures (for all Membrane Types)
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
Have Questions?
Catalog NumberWidth (Metric)Color
FIS08-671-5250 mmRed
Catalog number FIS08-671-52
Price (SGD)
335.00
Pack
Width (Metric):
50 mm
Color:
Red
Price (SGD)
335.00
Pack
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
  • Universal design with gentle sealing ridges, will not damage thicker walled tubing
  • Can be used with all membrane types: Standard RC, Biotech RC and Biotech CE
  • Easy to open and reseal
  • Color options for sample identification
  • Made of polyamide 6.6 (nylon)
  • Operating temperature: 0° to 90°C
  • Sterilizable by gamma irradiation or ethylene oxide (not autoclavable)
Specifications
Product TypeDialysis Tubing Closure
MembraneCellulose Ester, Regenerated Cellulose
MaterialNylon
Width (Metric)50 mm
AutoclavableNot Autoclavable
ColorRed
SterilitySterile
Temperature Range (English)32°F to 194°F
Temperature Range (Metric)0°C to 90°C
Quantity10 Pk.
For Use With (Application)Universal closures for use with all tubing membrane types
Unit SizePack