Spectra/Por™ 1 RC Dialysis Membrane Tubing 6000 to 8000 Dalton MWCO
Actual product may vary
Spectrum
Spectra/Por™ 1 RC Dialysis Membrane Tubing 6000 to 8000 Dalton MWCO
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Length (Metric)
FIS08-670F76 mm15 m
FIS08-670C25.5 mm30.5 m
FIS08-670E64 mm15 m
FIS08-670-36.4 mm15.2 m
Catalog number FIS08-670F
Price (SGD)
1,509.00
Each
Diameter (Metric):
76 mm
Length (Metric):
15 m
Price (SGD)
1,509.00
Each
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Supplied as flat-width tubing or flat sheets that can be folded to fit various funnel-type dialysis apparatus. Pure regenerated natural cellulose membranes with 1000 to 50,000 molecular weight cutoffs.
  • Transparent and flexible
  • Low molecular weight compounds in aqueous solutions diffuse through membrane
  • Higher molecular weights, such as proteins and bacteria, are retained by membrane
  • Glycerol preserves membrane and must be removed before use
  • Flat sheets and discs available for use with dialysis apparatus
Do not tie knots in tubing; use closures
Specifications
Product TypeSpectra/Por Dialysis Tubing
MWCO6 to 8 kDa
MaterialRegenerated Cellulose
FormatTubing
Diameter (Metric)76 mm
Diameter (English)3 in.
Length (English)49.2 ft.
Length (Metric)15 m
Temperature Range (English)39.2°F to 249.8°F
Temperature Range (Metric)4°C to 121°C
QuantityEach
For Use With (Application)Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Unit SizeEach