Spectra/Por™ 6 Pre-wetted Standard RC Dialysis Tubing, 1-50 kD MWCO
Actual product may vary
Spectrum
Spectra/Por™ 6 Pre-wetted Standard RC Dialysis Tubing, 1-50 kD MWCO
Clear, flexible and sturdy; Standard Grade RC (regenerated cellulose) membrane conveniently pre-equilibrated in water, eliminating the need to soak to remove glycerin and hydrate the membrane
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)MWCO
FIS08-670-19C21.6 mm50 kDa
FIS08-670-12C24 mm1 kDa
FKS1143586924 mm1 kDa
FIS08-670-12D29 mm1 kDa
FIS08-670-13C24 mm2 kDa
FIS08-670-14A11.5 mm3.5 kDa
FIS08-670-15B20.4 mm8 kDa
FIS08-670-16C20.4 mm10 kDa
FIS08-670-17C20.4 mm15 kDa
FIS08-670-19A7.5 mm50 kDa
FIS08-670-18C21.6 mm25 kDa
FIS08-670-14B29 mm3.5 kDa
Catalog number FIS08-670-19C
Price (SGD)
1,362.00
Each
Diameter (Metric):
21.6 mm
MWCO:
50 kDa
Price (SGD)
1,362.00
Each
Clear, flexible and sturdy; Standard Grade RC (regenerated cellulose) membrane conveniently pre-equilibrated in water, eliminating the need to soak to remove glycerin and hydrate the membrane
This ready-to-use membrane is packaged wet in a preservative solution of 0.05% sodium azide. Simply rinse away the preservative solution with DI water, load the sample and dialyze.
  • Pre-wetted & ready-to-use, no glycerin removal required
  • 8 MWCO's ranging from 1 kD - 50 kD
  • Good chemical compatibility with dilute strong acids and bases, conc. weak acids & bases, many alcohols and some organics
  • Standard RC can tolerate pH 2 - 12 and temp 4-121°C.
  • Ideal for sample prep, desalting, buffer exchange, pH change, protein purification, etc.
  • Economic 10 meters/pkg
Do not tie knots in tubing; use closures
Specifications
Product TypeSpectra/Por 6 Dialysis Tubing
MWCO50 kDa
MaterialRegenerated Cellulose
Diameter (Metric)21.6 mm
Diameter (English)0.85 in.
Length (English)33 ft.
Length (Metric)10 m
FormulationPackaged wet in 0.05% sodium azide
Temperature Range (English)39.2°F to 249.8°F
Temperature Range (Metric)4°C to 121°C
QuantityEach
For Use With (Application)Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Unit SizeEach