Spectrum™ Dialysis Tubing Closures: Spectra/Por™ Standard Type
Actual product may vary
Spectrum
Spectrum™ Dialysis Tubing Closures: Spectra/Por™ Standard Type
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWidth (Metric)Color
FIS08-670-11A35 mmOrange
FIS08-801-3635 mmBlue
FIS08-670-11AA12 mmOrange
FIS08-750-24B23 mmOrange
FIS08-750-24C55 mmOrange
FIS08-671-8755 mmGreen
Catalog number FIS08-670-11A
Price (SGD)
421.00
Pack
Width (Metric):
35 mm
Color:
Orange
Price (SGD)
421.00
Pack
Consist of a single-piece, hinged clamp with sealing ridges that securely lock over the dialysis tubing to provide a leak-proof seal
Spectra/Por Standard Closure Type:
  • Made from polypropylene to aid in sample buoyancy
  • Maintains sample vertical orientation when used with a Spectra/Por Weighted Closure.
  • 5 sealing width sizes: 12mm, 23mm, 35mm, 55mm and 75mm
  • 4 colors available for easier sample identification
  • Autoclavable
  • 10/pkg; Use with RC dialysis tubing only. Not for use with CE tubing.
Specifications
Product TypeDialysis Tubing Closure
MembraneRegenerated Cellulose
MaterialPolypropylene
Width (Metric)35 mm
AutoclavableAutoclavable
ColorOrange
Temperature Range (English)32°F to 250°F
Temperature Range (Metric)0°C to 121°C
QuantityEach
For Use With (Application)For sample buoyancy and vertical orientation, Spectrum laboratories, use with RC only
Unit SizePack