Search Fisher Scientific
| Catalog Number | Height (Metric) | Color | No. of Shelves | Volume (English) |
|---|---|---|---|---|
| FIS08-647-657 | 41.7 cm | Clear | 3 | 1.58 cu. ft. |
| FIS08-647-676 | 51.8 cm | Blue with Clear Door | 3 | 1.9 cu. ft. |
| FIS08-647-54 | 51.8 cm | Clear | 3 | 1.9 cu. ft. |
| FIS08-647-851 | 31.4 cm | Amber | 2 | 1.17 cu. ft. |
| FKS11842742 | 31.4 cm | Clear | 2 | 1.17 cu. ft. |
Constructed of Durastar™ co-polyester. Bel-Art™ Secador™ Standard Desiccator Cabinets: Vertical Models block 99% of UV light and are resistant to staining, crazing, and chemical attack.