Porcelain Desiccator Plates for Crucibles
Porcelain Desiccator Plates for Crucibles
Actual product may vary
Porcelain Desiccator Plates for Crucibles
With 30mm diameter holes for crucibles
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (English) OuterDiameter (Metric) OuterProduct TypeHolds
FIS08-636D5.74 in.14.6 cmDesiccator Plate w/Feet7
FIS08-636A3.74 in.9.5 cmDesiccator Plate w/Feet3
FIS08-636B4.52 in.11.5 cmDesiccator Plate w/Feet4
Catalog number FIS08-636D
Price (SGD)
-
Diameter (English) Outer:
5.74 in.
Diameter (Metric) Outer:
14.6 cm
Product Type:
Desiccator Plate w/Feet
Holds:
7
  • With or without three 16mm feet to support plate
  • Glazed on top
Specifications
Diameter (Metric) Outer14.6 cm
Product TypeDesiccator Plate w/Feet
MaterialPorcelain
Holds7
IncludesThree 16mm feet
Diameter (English) Outer5.74 in.
Unit SizeEA