MIC Test Kit, Petroleum Microbiology Media: Sulfate Reducing Bacteria Media
Actual product may vary
MIC Test Kit, Petroleum Microbiology Media: Sulfate Reducing Bacteria Media
MIC Test Kit, Petroleum Microbiology Media: Sulfate Reducing Bacteria Media
MIC Test Kit, Petroleum Microbiology Media: Sulfate Reducing Bacteria Media
MIC Test Kit, Petroleum Microbiology Media: Sulfate Reducing Bacteria Media
MIC Test Kit, Petroleum Microbiology Media: Sulfate Reducing Bacteria Media
Microbiology growth media products
Have Questions?
Catalog NumberQuantityMedia TypePackaging Type
FIS08-629-001A12 VialsSRB, 1% salinityVial
Catalog number FIS08-629-001A
Price (SGD)
-
Quantity:
12 Vials
Media Type:
SRB, 1% salinity
Packaging Type:
Vial
A range of bacterial growth media and accessories for use in detecting and quantifying microbiologically influenced corrosion (MIC). Media products are available as boxes of 12 vials, cases of 288 vials, and in the single tube quick-test format.
Sulfate Reducing Bacteria (SRB) contribute to MIC by producing hydrogen sulfide, which is a corrosive and toxic gas that has the odor of rotten eggs. SRB can also sour oil and gas reservoirs, create precipitates that plug openings and discolor water to black.
Sulfate Reducing Bacteria Media
  • SRB: for quantification of sulfate reducing bacteria (Modified Postgates B Medium)
  • SRB-Q: quick format single tube test for SRB
  • SRB2: specially formulated clear SRB media
  • SRB-Q media provides a convenient way to estimate the concentration of SRB in water samples by changing the clarity of the color of the media from white to black
Specifications
Certifications/ComplianceNACE, TM0194-94
ColorClear
Manufacturing OriginUnited States
Media TypeSRB, 1% salinity
Product TypeMIC Test Kit
For Use With (Application)Cultivation of Sulfate reducing bacteria, MIC testing
Packaging TypeVial
Quantity12 Vials
Unit SizeEach