Spectra/Por Biotech-Grade RC Dialysis Membrane Tubing
Actual product may vary
Spectrum
Spectra/Por Biotech-Grade RC Dialysis Membrane Tubing
A premium dialysis membrane that provides superior molecular selectivity than Standard RC and superior tolerance than Biotech CE
Have Questions?
Catalog NumberDiameter (Metric)MWCO
FIS08-607-0676.4 mm20 kDa
Catalog number FIS08-607-067
Price (SGD)
1,048.00
Each
Diameter (Metric):
6.4 mm
MWCO:
20 kDa
Price (SGD)
1,048.00
Each
  • Crafted from a regeneration process that yields superior temperature tolerances and chemical compatibilities with the same high purity and selectivity as Biotech CE
  • Membrane can tolerate concentrated-weak acids and bases, dilute-strong acids and bases, most alcohols and some mild or dilute organic solvents
  • Rigidly controlled porosity for better selectivity and separations
  • Ultrapure, does not interfere with molecular activity and function
  • 3 MWCOs and 2 tubing flat-widths
  • Packaged dry with glycerin (soak 15 min. in water)
  • Use with pH 2 to 12 and temp 4 to 60°C
Specifications
Product TypeSpectra/Por Dialysis Tubing
MWCO20 kDa
MaterialRegenerated Cellulose
Diameter (Metric)6.4 mm
Diameter (English)0.25 in.
Length (English)16.4 ft.
Length (Metric)5 m
FormulationPackaged dry with glycerin (soak 15 min. in water)
Temperature Range (English)39.2°F to 140°F
Temperature Range (Metric)4°C to 60°C
QuantityEach
For Use With (Application)Frequent dialysis applications, desalting, purification, sample preparation
Unit SizeEach