Class B Cylinders with Single Amber Metric Scale
Class B Cylinders with Single Amber Metric Scale
Actual product may vary
Class B Cylinders with Single Amber Metric Scale
Calibrated To Deliver
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)GraduationsTolerance MetricSubdivision
FIS08-570H1000 mL10 mL±6 mL10mL
FIS08-570B10 mL0.2 mL±0.2 mL0.2mL
FIS08-570A5 mL0.1 mL±0.1 mL0.1mL
FIS08-570C25 mL0.5 mL±0.34 mL0.5mL
FIS08-570F250 mL2 mL±2 mL2mL
FIS08-570E100 mL1 mL±1 mL1mL
FIS08-570D50 mL1 mL±0.5 mL1mL
FIS08-570G500 mL5 mL±4 mL5mL
Catalog number FIS08-570H
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1000 mL
Graduations:
10 mL
Tolerance Metric:
±6 mL
Subdivision:
10mL
Calibrated To Deliver
  • Ideal for use with light-colored liquids
  • Dark amber graduations provide contrast for easy readings
  • Glass cylinders with heavy round bases and pouring spouts
  • Accuracy is assured even at lowest graduations; first 10% of volume is not graduated
Specifications
Base TypeRound
CalibrationTo Deliver
Scale ColorAmber
ShapeRound
Certifications/ComplianceASTM E1272
ClassB
Height (English)18.1 in.
Height (Metric)460 mm
Tolerance Metric±6 mL
TypeGraduated Cylinder
MaterialBorosilicate Glass
Capacity (Metric)1000 mL
For Use With (Application)Ideal for use with Light Colored Liquids, Amber Graduations Provide Contrast for Easy Readings, Heavy Round Bases and Pouring Spouts
ScaleSingle
Graduations10 mL
Subdivision10mL
Unit SizeEA