PYREX™ Class B Mixing Cylinders with Standard Taper Stopper, White Graduations
PYREX™ Class B Mixing Cylinders with Standard Taper Stopper, White Graduations
Actual product may vary
PYREX™ Class B Mixing Cylinders with Standard Taper Stopper, White Graduations
Calibrated To Contain
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduations
FIS08-566G1 L10 mL
FIS08-566E250 mL2 mL
FIS08-566F500 mL5 mL
Catalog number FIS08-566G
Price (SGD)
543.00
CS
Add to cart
Capacity (Metric):
1 L
Graduations:
10 mL
Price (SGD)
543.00
CS
Add to cart
Calibrated 'to contain' in accordance with ASTM E-1272 and ASTM E-542. Useful as a mixing cylinder. Each features the PYREX stopper. Graduations are in durable white enamel. See Cat. No. 3002 for the equivalent cylinder supplied with a Corning test certificate.
For stopper only, see 7650.
Specifications
Base TypeHexagonal
CalibrationTo Contain
Scale ColorWhite
ShapeRound
SterilityNon-sterile
TypeGraduated Mixing Cylinder
Tolerance±6 mL
MaterialBorosilicate Glass
Certifications/ComplianceASTM
ClassB
Diameter (English) Outside2.51 in.
Capacity (Metric)1 L
Diameter (Metric) Outside64 mm
For Use With (Application)Useful as a Mixing Cylinder
Height (English)20.4 in.
Height (Metric)520 mm
IncludesStopper
ScaleSingle
Stopper Number32
Stopper MaterialGlass
Stopper StyleHollow
Tolerance (Metric)±6 mL
Closure MaterialGlass
Graduations10 mL
AutoclavableAutoclavable
Subdivision10 mL
Unit SizeCS