Class B Mixing Cylinders
Class B Mixing Cylinders
Actual product may vary
Class B Mixing Cylinders
Calibrated To Contain
Have Questions?
Catalog NumberCapacity (Metric)GraduationsLimit of Error
FIS08-565E250 mL2 mL+/- 1.4mL
Catalog number FIS08-565E
Price (SGD)
184.00
EA
Add to cart
Capacity (Metric):
250 mL
Graduations:
2 mL
Limit of Error:
+/- 1.4mL
Price (SGD)
184.00
EA
Add to cart
Calibrated To Contain
  • Double metric scale
  • White-enamel graduations
  • Ground-glass pennyhead stoppers First 10% of volume not graduated
  • Autoclavable up to 121°C, 10- to 15-minute intervals recommended
Specifications
CalibrationTo Contain
Scale ColorWhite
TypeGraduated Mixing Cylinder
Tolerance±1.4 mL
MaterialBorosilicate Glass
ClassB
Capacity (Metric)250 mL
ScaleDual
Stopper NumberUse with a #27 Stopper
Stopper StylePennyhead
Tolerance (Metric)±1.4mL
Closure MaterialGlass
Graduations2 mL
AutoclavableAutoclavable
Limit of Error+/- 1.4mL
Subdivision2mL
Unit SizeEA