Serialized Class A Cylinders with Certificate of Traceability
Serialized Class A Cylinders with Certificate of Traceability
Actual product may vary
Serialized Class A Cylinders with Certificate of Traceability
Calibrated To Contain
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)GraduationsLimit of Error
FIS08-559H2000 mL20 mL+/- 6.0mL
FIS08-559A10 mL0.1 mL+/- 0.10mL
FIS08-559D100 mL1 mL+/- 0.50mL
FIS08-559F500 mL5 mL+/- 2.0mL
FIS08-559G1000 mL10 mL+/- 3.0mL
Catalog number FIS08-559H
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
2000 mL
Graduations:
20 mL
Limit of Error:
+/- 6.0mL
Calibrated To Contain
  • Serialized Class A
  • With traceability
  • Borosilicate glass
  • White scale
Specifications
CalibrationTo Contain
Scale ColorWhite
ShapeRound
ClassA
Height (English)20.4 in.
Height (Metric)520 mm
Tolerance Metric±6 mL
TypeGraduated Cylinder
MaterialBorosilicate Glass
Capacity (Metric)2000 mL
IncludesTraceable NIST Certificate
ScaleWhite scale
Graduations20 mL
AutoclavableAutoclavable
Limit of Error+/- 6.0mL
Subdivision20mL
Unit SizeEA