Check Order Status
Serialized Class A Graduated Cylinders with Certificate of Calibration
Actual product may vary
Serialized Class A Graduated Cylinders with Certificate of Calibration
Calibrated
To Contain
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Graduations
FIS08-559FC
500 mL
5 mL
FIS08-559AC
10 mL
0.1 mL
FIS08-559DC
100 mL
1 mL
3 Options
Catalog number
FIS08-559FC
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
500 mL
Graduations:
5 mL
Calibrated
To Contain
.
White scale graduations
Borosilicate glass
Case of 1
Specifications
Calibration
To Contain
Description
Cs. of 1
Scale Color
White
Shape
Round
Class
A
Height (English)
14.7 in.
Height (Metric)
375 mm
Tolerance Metric
±2 mL
Type
Graduated Cylinder
Material
Glass
Capacity (Metric)
500 mL
Includes
Traceable NIST Certificate
Scale
White scale
Graduations
5 mL
Autoclavable
Autoclavable
Limit of Error
±2.00 mL
Subdivision
5mL