PYREX™ Cylinder for Gas Oil Distillations
PYREX™ Cylinder for Gas Oil Distillations
Actual product may vary
PYREX™ Cylinder for Gas Oil Distillations
Calibrated To Contain in percent
Have Questions?
Catalog NumberCapacity (Metric)
FIS08-554200 mL
Catalog number FIS08-554
Price (SGD)
3,156.00
EA
Add to cart
Capacity (Metric):
200 mL
Price (SGD)
3,156.00
EA
Add to cart
These 200mL PYREX™ graduated cylinders are calibrated “to contain” (TC) in percent. 200mL is equal to 100% in 1% divisions. The scale is in durable white enamel. This cylinder may be used to measure the distillate in ASTM method D-86 “Distillation of Gas Oil and Similar Distillate Oil.” Furnished with bumper guard. For replacement bumper guards, see No. 3066.
Specifications
Base TypeHexagonal
CalibrationTo Contain
IncludesBumper Guard
Scale ColorWhite
ShapeRound
Tolerance±1.4 mL
MaterialBorosilicate Glass
Certifications/ComplianceASTM E438
Diameter (English) Outside1.6 in.
Diameter (Metric) Outside41 mm
For Use With (Application)Used to Measure the Distillate in Astm Method D-86 Distillation of Gas Oil and Similar Distillate Oil
Capacity (Metric)200 mL
For Use With (Equipment)Bumper Guard
GraduatedYes
Height (English)10 in.
Height (Metric)257 mm
ScaleWhite enamel scale
Tolerance (Metric)1.4 mL
Product TypeGraduated Distillation Cylinder
Graduations2 mL
AutoclavableAutoclavable
Subdivision2 mL