PYREX™ Cylinders with Single White-Enamel Metric Graduations, TC
PYREX™ Cylinders with Single White-Enamel Metric Graduations, TC
Actual product may vary
PYREX™ Cylinders with Single White-Enamel Metric Graduations, TC
Calibrated To Contain
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduations
FIS08-552G500 mL5 mL
FIS08-552D50 mL1 mL
FIS08-552F250 mL2 mL
Catalog number FIS08-552G
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
500 mL
Graduations:
5 mL
Calibrated 'to contain,' with white enamel graduations. The 100 mL capacity is in accordance with ASTM E-133 and E-1272 and is used in ASTM tests D-86, D-216 and D-447. The 10 mL size has a funnel top. Bumper guards are supplied with 25 mL through 2000 mL sizes inclusive.
Specifications
Base TypeHexagonal
CalibrationTo Contain
Scale ColorWhite
ShapeRound
SterilityNon-sterile
Certifications/ComplianceASTM D216, ASTM D447, ASTM D86, ASTM E1272, ASTM E133
ClassB
Height (English)15.4 in.
Height (Metric)392 mm
Tolerance Metric±4 mL
TypeGraduated Cylinder
MaterialBorosilicate Glass
Diameter (English) Outside1.9 in.
Diameter (Metric) Outside50 mm
Capacity (Metric)500 mL
IncludesBumper Guard
ScaleSingle
Graduations5 mL
AutoclavableAutoclavable
Subdivision5 mL
Unit SizeEA