Standard Class B Graduated Cylinders
Standard Class B Graduated Cylinders
Actual product may vary
Standard Class B Graduated Cylinders
Standard Class B Graduated Cylinders
Standard Class B Graduated Cylinders
Standard Class B Graduated Cylinders
Standard Class B Graduated Cylinders
Calibrated To Deliver
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)GraduationsTolerance MetricSubdivision
FIS08-550H1000 mL10 mL±6 mL10mL
FIS08-550B10 mL0.2 mL±0.2 mL0.2mL
FIS08-550D50 mL1 mL±0.5 mL1mL
FIS08-550A5 mL0.1 mL±0.1 mL0.1mL
FIS08-550F250 mL2 mL±2 mL2mL
FIS08-550C25 mL0.5 mL±0.34 mL0.5mL
FIS08-550G500 mL5 mL±4 mL5mL
FIS08-550E100 mL1 mL±1 mL1mL
Catalog number FIS08-550H
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1000 mL
Graduations:
10 mL
Tolerance Metric:
±6 mL
Subdivision:
10mL
Calibrated To Deliver
  • White-ceramic graduations
  • Heavy round bases
  • Handy pouring spouts
  • Accuracy is assured even at the lowest graduations; first 10% volume is not graduated
Specifications
Base TypeRound
CalibrationTo Deliver
Scale ColorWhite
ShapeRound
ClassB
Height (English)18.1 in.
Height (Metric)460 mm
Tolerance Metric±6 mL
TypeGraduated Cylinder
MaterialBorosilicate Glass
Capacity (Metric)1000 mL
For Use With (Application)Accuracy assured even at lowest graduations
ScaleWhite ceramic graduations
Graduations10 mL
Subdivision10mL
Unit SizeEA