PYREX™ Hydrometer-Jar-Cylinders
PYREX™ Hydrometer-Jar-Cylinders
Actual product may vary
PYREX™ Hydrometer-Jar-Cylinders
Ungraduated
Have Questions?
Catalog NumberCapacity (Metric)
FIS08-532C500 mL
Catalog number FIS08-532C
Price (SGD)
129.00
EA
Add to cart
Capacity (Metric):
500 mL
Price (SGD)
129.00
EA
Add to cart
Ungraduated
Breakage is minimized by the heavy wall construction. The large hexagonal base, sealed to the cylinder body, increases stability. Meets ASTM specifications D-287 and E-100.
Specifications
ShapeRound
Base TypeHexagonal
Height (English)14.76 in.
Height (Metric)39 cm
TypeHydrometer Cylinder
Diameter (English)1.96 in.
Diameter (Metric)5 cm
Capacity (Metric)500 mL
MaterialBorosilicate Glass
No. per Pack1/Pk.
No. per Case12/Cs.
Certifications/ComplianceASTM D287, ASTM E100