Porcelain Combustion Boats
Actual product may vary
CoorsTek
Porcelain Combustion Boats
AD-998 High-Alumina labware
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (English)Length (English)Width (English)
FIS08-226-13G50 mL0.82 in.5.3 in.1.06 in.
FIS08-226-13B3 mL0.35 in.1.96 in.0.47 in.
FIS08-226-13C5 mL0.39 in.2.75 in.0.55 in.
FIS08-226-13E15 mL0.51 in.3.93 in.0.78 in.
FIS08-226-13F20 mL0.57 in.4.13 in.0.86 in.
FIS08-226-13D10 mL0.45 in.3.54 in.0.66 in.
Catalog number FIS08-226-13G
Price (SGD)
550.00
Each
Capacity (Metric):
50 mL
Height (English):
0.82 in.
Length (English):
5.3 in.
Width (English):
1.06 in.
Price (SGD)
550.00
Each
Specifications
Bottom ShapeFlat
Capacity (Metric)50 mL
MaterialAluminum, Porcelain
ShapeBoat
TypeCombustion Boat
For Use With (Application)For refractory metals such as molybdenum, platinum, rhodium, tungsten, tantalum and iridium
Dimensions (L x W x H)5.3 x 1.06 x 0.82 in. (137 x 27 x 21 mm)
Length (English)5.3 in.
Length (Metric)137 mm
Width (English)1.06 in.
Width (Metric)27 mm
Height (Metric)21 mm
Height (English)0.82 in.
Unit SizeEach