Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Specialty Lab Equipment, Instruments, and Apparatuses
▸
Combustion Analyzers and Products
▸
Combustion Boats
Actual product may vary
CoorsTek
Combustion Boats
Have Questions?
Catalog Number
Color
FIS08-226-13A
White
Catalog number
FIS08-226-13A
Price (SGD)
195.00
Each
Add to cart
Color:
White
Price (SGD)
195.00
Each
Add to cart
Specifications
Type
Combustion Boat
Color
White
Dimensions (L x W x H)
30 x 8 x 7 mm
Height (Metric)
7 mm
Length (Metric)
30 mm
Material
High Alumina
Width (Metric)
8 mm