Search Fisher Scientific
| Catalog Number | Capacity (Metric) | Diameter (Metric) Inner | Height (Metric) |
|---|---|---|---|
| FIS08-226-12K | 180 mL | 54mm | 91mm |
Highly resistant to chemical attack, this alumina-ceramic crucible is especially useful to chemists, metallurgists and others involved in high-temperature work
Suitable for glass melting, including borosilicate glass. Recommended for use with refractory metals such as molybdenum, platinum, rhodium, tungsten, tantalum and iridium