Porcelain Coal Moisture Testing Crucibles
Porcelain Coal Moisture Testing Crucibles
Actual product may vary
Porcelain Coal Moisture Testing Crucibles
Withstand temperatures up to 1150°C (2102°F), not to exceed 200°C/hour
Have Questions?
Catalog NumberCapacity (Metric)
FIS08-103B25 mL
Catalog number FIS08-103B
Price (SGD)
-
Capacity (Metric):
25 mL
  • Porcelain with rounded bottom
  • Glazed inside and out except on rim
  • 20mL size recommended for ASTM™ D 3176
  • Capsule form; without covers
Specifications
MaterialPorcelain
Capacity (Metric)25 mL
Unit SizeEA