MASTERBLOCK™ 96 Deep Well Conical Bottom 0.5mL Storage Plate
MASTERBLOCK™ 96 Deep Well Conical Bottom 0.5mL Storage Plate
Actual product may vary
Fisherbrand
MASTERBLOCK™ 96 Deep Well Conical Bottom 0.5mL Storage Plate
Suitable for cultivating bacteria or yeasts. Greiner Bio-One MASTERBLOCK™ 96 Deep Well Conical Bottom 0.5mL Storage Plate is also suitable for storing sample material.
Have Questions?
Catalog NumberSterility
FIS07000169Sterile
Catalog number FIS07000169
Price (SGD)
1,638.00
CS
Add to cart
Sterility:
Sterile
Price (SGD)
1,638.00
CS
Add to cart
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Sealable with adhesive films and heat sealer
  • High temperature resistance
  • High chemical resistance
  • A
Specifications
FormatStandard
Array8 x 12
ColorNatural
MaterialPolypropylene
No. of Wells96
SterilitySterile
Total Volume (Metric) Well0.5 mL
Well ShapeConical Bottom (V-well)
No. per Case80
No. per Pack1
Quantity80/Cs.
Unit SizeCS