Check Order Status
High-Form Porcelain Crucibles
Actual product may vary
High-Form Porcelain Crucibles
Withstand temperatures up to 1150°C (2102°F), not to exceed 200°C/hour
Have Questions?
Change view
Catalog Number
Capacity (Metric)
FIS07-965G
80 mL
FIS07-965-1
250 mL
2 Options
Catalog number
FIS07-965G
Price (SGD)
-
Capacity (Metric):
80 mL
Glazed inside and out except for outside bottom surface and rim
Covers sold separately
Specifications
Material
Porcelain
Capacity (Metric)
80 mL
Diameter (Metric) Outer
60 mm
Height (Metric)
43 mm