Rilmenidine hemifumarate
Rilmenidine hemifumarate
Actual product may vary
Rilmenidine hemifumarate
Rilmenidine hemifumarate
Rilmenidine hemifumarate
α2 agonist. Also I1 ligand
Have Questions?
Catalog NumberQuantity
FIS07-9055 mg
Catalog number FIS07-905
Price (SGD)
32.00
EA
Add to cart
Quantity:
5 mg
Price (SGD)
32.00
EA
Add to cart
I1-imidazoline binding site selective ligand and α2-adrenoceptor agonist. Possesses greater I1 vs α2 selectivity than the prototypical compound, clonidine. Also thought to enhance autophagy; shown to increase LC3-II levels in PC12 cells.
Specifications
Chemical Name or Material2-[N-(Dicyclopropylmethyl)amino]oxazoline hemifumarate
CAS207572-68-7
Assay Percent Range>99%
Recommended StorageStore at -20°C
Quantity5 mg
Molecular FormulaC10H16N2O.½C4H4O4
Formula WeightObserved MW: 238.29
Purity0.98
Unit SizeEA