Cork Supports
Cork Supports
Actual product may vary
Cork Supports
Durable, compressed cork lab rings
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (English) OuterDiameter (Metric) InnerDiameter (Metric) OuterFor Use With (Equipment)
FIS07-835D8.26 in.15 cm21 cmConcave recesses support dishes and 12000mL round bottom flasks
FIS07-835A4.33 in.6 cm11 cmConcave recesses support dishes and 200 to 500mL round bottom flasks
FIS07-835AA3.14 in.3 cm8 cmConcave recesses support dishes and 10 to 100mL round bottom flasks
FIS07-835B5.15 in.9 cm14 cmConcave recesses support dishes and 1000 to 3000mL round bottom flasks
FIS07-835C6.69 in.12 cm17 cmConcave recesses support dishes and 5000mL round bottom flasks
Catalog number FIS07-835D
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (English) Outer:
8.26 in.
Diameter (Metric) Inner:
15 cm
Diameter (Metric) Outer:
21 cm
For Use With (Equipment):
Concave recesses support dishes and 12000mL round bottom flasks
Durable, compressed cork lab rings
  • Concave recesses support dishes and round-bottom flasks
  • Durable
Specifications
Product TypeCork Ring Stand
AutoclavableNot Autoclavable
MaterialCork
ShapeRound
For Use With (Equipment)Concave recesses support dishes and 12000mL round bottom flasks
Height (Metric)3 cm
Height (English)1.18 in.
Diameter (English) Inner5.9 in.
Diameter (English) Outer8.26 in.
Diameter (Metric) Outer21 cm
Diameter (Metric) Inner15 cm
Unit SizeEA