Cork Stoppers - Superior Grade
Cork Stoppers - Superior Grade
Actual product may vary
Cork Stoppers - Superior Grade
Corks are cut from high-quality wood and are selected to meet superior standards
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric) BottomStopper NumberQuantity
FIS07-791F2.22 cm1.34 cm5100/Pk.
FIS07-791A1.27 cm0.71 cm0100/Pk.
FIS07-791B1.58 cm0.83 cm1100/Pk.
Catalog number FIS07-791F
Price (SGD)
19.00
PK
Add to cart
Length (Metric):
2.22 cm
Diameter (Metric) Bottom:
1.34 cm
Stopper Number:
5
Quantity:
100/Pk.
Price (SGD)
19.00
PK
Add to cart
Corks are cut from high-quality wood and are selected to meet superior standards
Specifications
For Use With (Application)Ideal for sealing bottles
GradeSuperior
Length (Metric)2.22 cm
Quantity100/Pk.
Size5
ColorNatural
Diameter (Metric) Bottom1.34 cm
Diameter (Metric) Top1.75 cm
MaterialWood
Product TypeCork Stopper
Stopper Number5
Unit SizePK