PYREX™ Graham Condensers with Standard Taper Inner and Outer Joints
PYREX™ Graham Condensers with Standard Taper Inner and Outer Joints
Actual product may vary
PYREX™ Graham Condensers with Standard Taper Inner and Outer Joints
For use in vacuum distillation
Have Questions?
Catalog NumberHeight (English)Length (English) JacketStandard Taper Joint No
FIS07-728A17.71 in.11.81 in.24/40
Catalog number FIS07-728A
Price (SGD)
793.00
EA
Add to cart
Height (English):
17.71 in.
Length (English) Jacket:
11.81 in.
Standard Taper Joint No:
24/40
Price (SGD)
793.00
EA
Add to cart
For use in vacuum distillation
This condenser has a high surface area per unit length of jacket. Primarily used in vacuum ­distillations. With the outer joint at the top and the inner standard taper joint at the bottom with a drip tip. The tubulation O.D. is approximately 10 mm.
Specifications
Diameter (English) Outer Jacket1.61 in.
Diameter (English) Outer Tubulation0.39 in.
Diameter (Metric) Outer Jacket41 mm
Diameter (Metric) Outer Tubulation10 mm
For Use With (Application)For vacuum distillations
Height (English)17.71 in.
Height (Metric)450 mm
Length (English) Jacket11.81 in.
Length (Metric) Jacket300 mm
Standard Taper Joint No24/40
MaterialBorosilicate Glass
TypeGraham Condenser
Unit SizeEA