ThermalTray™ Thermo-conductive Platforms
Actual product may vary
ThermalTray™ Thermo-conductive Platforms
Support Corning CoolRack™ and CoolSink™ sample modules in liquid temperature sources such as melting ice, water baths, and liquid nitrogen.
Have Questions?
Catalog NumberFor Use With (Equipment)Type
FIS07-210-0739L ice pan w/ liquid nitrogenSLP, Slim, Low Profile
Catalog number FIS07-210-073
Price (SGD)
-
For Use With (Equipment):
9L ice pan w/ liquid nitrogen
Type:
SLP, Slim, Low Profile
Corning ThermalTray™ thermo-conductive platforms support CoolRack™ and CoolSink™ sample modules in liquid temperature sources such as melting ice, water baths and liquid nitrogen. Made of the same highly conductive alloy as CoolRack and CoolSink modules, ThermalTray platforms conduct the source temperature to the CoolRack or CoolSink and, ultimately, to your samples.
  • Stable, sturdy design makes them ideal for processing temperature-sensitive samples in melted ice baths or liquid nitrogen.
  • All modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
Specifications
DescriptionThermalTray Thermo-conductive platform
For Use With (Equipment)9L ice pan w/ liquid nitrogen
TypeSLP, Slim, Low Profile
Unit SizeCase