Costar™ 12-well Clear TC-treated Multiple Well Plates, Individually Wrapped, Sterile
Costar™ 12-well Clear TC-treated Multiple Well Plates, Individually Wrapped, Sterile
Actual product may vary
Costar™ 12-well Clear TC-treated Multiple Well Plates, Individually Wrapped, Sterile
Tissue culture-treated plates for superior cell attachment. Corning™ Costar™ Flat Bottom Cell Culture Microplates provide an excellent device for diagnostic testing when cell adherence is essential.
Have Questions?
Catalog NumberDiameter (Metric)
FIS07-200-822.26 cm
Catalog number FIS07-200-82
Price (SGD)
748.00
CS
Add to cart
Diameter (Metric):
2.26 cm
Price (SGD)
748.00
CS
Add to cart
  • Individual alphanumerical codes for well identification, flat bottoms
  • Treated for optimal cell attachment (except where noted)
  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
  • Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
  • Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
  • Sterile
  • Nonpyrogenic
Specifications
Array3 x 4
Closure ColorClear
Closure IncludedYes
Closure MaterialPolystyrene
Bottom ShapeFlat
Capacity (Metric)1.14 mL (Well)
ColorClear
Culture Area3.8 cm2
MaterialPolystyrene
ShapeRectangular
SterilitySterile
Surface TreatmentCell Culture
TypeMulti-well Cell Culture Plate
Volume (Metric) Working1 mL
Well Count12
IncludesLid
No. per Case50/Cs.
No. per Pack1/Pk.
PackagingIndividually Wrapped
Quantity50/Cs.
Diameter (Metric)2.26 cm
Unit SizeCS