96-Well Solid Black or White Polystyrene Microplates
96-Well Solid Black or White Polystyrene Microplates
Actual product may vary
96-Well Solid Black or White Polystyrene Microplates
Manufactured from medical grade polystyrene for strength. Corning™ 96-Well Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorSurface Treatment
FIS07-200-590BlackNon-treated
FIS07-200-589WhiteNon-treated
Catalog number FIS07-200-590
Price (SGD)
1,677.00
CS
Add to cart
Color:
Black
Surface Treatment:
Non-treated
Price (SGD)
1,677.00
CS
Add to cart
  • Designed to reduce well-to-well cross-talk
  • White microplates enhance luminescent signals and have low background luminescence.
  • Black microplates have low background fluorescence and minimize light scattering.
  • Specifications
    ColorBlack
    MaterialPolystyrene
    No. of Wells96
    Volume (Metric) Working75 to 200 μL
    SterilityNon-sterile
    Surface TreatmentNon-treated
    Well ShapeFlat Bottom (F-well)
    For Use With (Application)Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions.
    Unit SizeCS